KEY465194313 Key Organics / BIONET

(2Z)-2-[4-(3-fluoropropoxy)phenyl]-3-[(4-methylphenyl)amino]prop-2-enenitrile

  • MDL number MFCD02185998
  • Cas Number : 1164536-11-1
  • Molecular Weight : 310.372

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68911

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-0165_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart