KEY465194319 Key Organics / BIONET

(3E)-3-{[(4-chlorophenyl)methoxy]imino}-2-[4-(3-fluoropropoxy)phenyl]propanenitrile

  • MDL number MFCD02186004
  • Cas Number : 477889-17-1
  • Molecular Weight : 360.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68919

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-0184_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart