KEY465194327 Key Organics / BIONET

4-benzoyl-N-[3-(dimethylamino)propyl]-1H-pyrrole-2-carboxamide

  • MDL number MFCD02571673
  • Cas Number : 477889-30-8
  • Molecular Weight : 299.374

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68936

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-0216_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart