KEY465194349 Key Organics / BIONET

2-(3,5-dimethyl-1H-pyrazol-1-yl)-5-nitropyridine

  • MDL number MFCD02102602
  • Cas Number : 76211-92-2
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69001

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-0638_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart