KEY465194366 Key Organics / BIONET

1,4-dimethyl (2E)-2-(1-{[(3,4-dimethoxyphenyl)methyl]amino}ethylidene)butanedioate

  • MDL number MFCD02186030
  • Cas Number : 477890-06-5
  • Molecular Weight : 337.372

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69050

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-0800_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart