KEY465194371 Key Organics / BIONET

N-[2-(1H-indol-3-yl)ethyl]-3-nitropyridin-2-amine

  • MDL number MFCD02186031
  • Cas Number : 18527-84-9
  • Molecular Weight : 282.303

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69060

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-0813_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart