KEY465194375 Key Organics / BIONET

methyl 2-[1-(3,5-dichlorophenyl)-5-hydroxy-3-methyl-1H-pyrazol-4-yl]acetate

  • MDL number MFCD02571693
  • Cas Number : 477711-21-0
  • Molecular Weight : 315.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69064

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-0817_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart