KEY465194381 Key Organics / BIONET

methyl 2-methyl-5-oxo-1-(2-phenylethyl)-4,5-dihydro-1H-pyrrole-3-carboxylate

  • MDL number MFCD01893900
  • Cas Number : 371144-81-9
  • Molecular Weight : 259.305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69083

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-0845_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart