KEY465194387 Key Organics / BIONET

[(E)-2-{1-[3,5-bis(trifluoromethyl)phenyl]-4-nitro-1H-1,2,3-triazol-5-yl}ethenyl]dimethylamine

  • MDL number MFCD02186106
  • Cas Number : 477890-37-2
  • Molecular Weight : 395.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69091

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-0856_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart