KEY465194410 Key Organics / BIONET

4-(2,4-dichlorophenyl)-7-thia-2,5-diazatricyclo[6.4.0.0^{2,6}]dodeca-1(8),3,5-triene-3-carbaldehyde

  • MDL number MFCD02187370
  • Cas Number : 478029-48-0
  • Molecular Weight : 351.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69172

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-1152_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart