KEY465194414 Key Organics / BIONET

4-(4-chlorophenyl)-7-thia-2,5-diazatricyclo[6.4.0.0^{2,6}]dodeca-1(8),3,5-triene-3-carbaldehyde

  • MDL number MFCD02187372
  • Cas Number : 478029-59-3
  • Molecular Weight : 316.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69186

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-1171_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart