KEY465194415 Key Organics / BIONET

3-(4-chlorophenyl)-3-hydroxy-1-{2-methylimidazo[1,2-a]pyridin-3-yl}propan-1-one

  • MDL number MFCD02186621
  • Cas Number : 478029-60-6
  • Molecular Weight : 314.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69187

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-1182_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart