KEY465194425 Key Organics / BIONET

2-[(4-methoxyphenyl)sulfanyl]-1-{2-methylimidazo[1,2-a]pyridin-3-yl}ethan-1-one

  • MDL number MFCD02186299
  • Cas Number : 478029-75-3
  • Molecular Weight : 312.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69207

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-1210_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart