KEY465194428 Key Organics / BIONET

(3Z)-3-[2-(2,4,6-trichlorophenyl)hydrazin-1-ylidene]-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD02154634
  • Cas Number : 298207-63-3
  • Molecular Weight : 340.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69212

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-1251_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart