KEY465194433 Key Organics / BIONET

2-[3-(4-chlorophenyl)-1,2,4-oxadiazol-5-yl]acetic acid

  • MDL number MFCD02102656
  • Cas Number : 478030-50-1
  • Molecular Weight : 238.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69302

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-1555_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart