KEY465194434 Key Organics / BIONET

1-[(3-nitrophenyl)methyl]-2-(trifluoromethyl)-1H-1,3-benzodiazole

  • MDL number MFCD02102658
  • Cas Number : 478030-52-3
  • Molecular Weight : 321.259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69304

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-1557_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart