KEY465194437 Key Organics / BIONET

2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]propanoic acid

  • MDL number MFCD02571586
  • Cas Number : 478030-59-0
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69314

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-1568_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart