KEY465194438 Key Organics / BIONET

1-(4-fluorophenyl)-2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]ethan-1-one

  • MDL number MFCD02571587
  • Cas Number : 478030-60-3
  • Molecular Weight : 322.263

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69315

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-1569_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart