KEY465194441 Key Organics / BIONET

1-(4-chlorobenzenesulfonyl)-N'-[(1E)-(furan-2-yl)methylidene]piperidine-4-carbohydrazide

  • MDL number MFCD02186319
  • Cas Number : 478030-78-3
  • Molecular Weight : 395.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69335

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-1596_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart