KEY465194442 Key Organics / BIONET

(2Z)-3-(dimethylamino)-2-(4-phenyl-1,3-thiazol-2-yl)prop-2-enenitrile

  • MDL number MFCD03012214
  • Cas Number : 691879-58-0
  • Molecular Weight : 255.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69343

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-0005_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart