KEY465194443 Key Organics / BIONET

3-(4-fluorophenyl)-5-methyl-6-phenylpyrazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD03305534
  • Cas Number : 861209-16-7
  • Molecular Weight : 318.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69355

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-0027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart