KEY465194463 Key Organics / BIONET

3-{[(2-chloro-6-fluorophenyl)methyl]sulfanyl}-1-phenyl-5H,6H,7H-cyclopenta[c]pyridine-4-carbonitrile

  • MDL number MFCD03012238
  • Cas Number : 439096-34-1
  • Molecular Weight : 394.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69411

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-0261_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart