KEY465194469 Key Organics / BIONET

methyl 2-{4-[(2E)-3-phenylprop-2-enoyl]-3,4-dihydro-2H-1,4-benzoxazin-3-yl}acetate

  • MDL number MFCD03305562
  • Cas Number : 861209-30-5
  • Molecular Weight : 337.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69423

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-0279_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart