KEY465194471 Key Organics / BIONET

3-(cyclopentylsulfanyl)-1-phenyl-5H,6H,7H-cyclopenta[c]pyridine-4-carbonitrile

  • MDL number MFCD03305563
  • Cas Number : 861209-31-6
  • Molecular Weight : 320.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69426

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0282_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart