KEY465194476 Key Organics / BIONET

4-{4-[(4-methoxyphenyl)methyl]-5-methyl-4H-1,2,4-triazol-3-yl}-1-methyl-1H-pyrazol-5-amine

  • MDL number MFCD03305567
  • Cas Number : 955976-91-7
  • Molecular Weight : 298.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69435

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0295_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart