KEY465194477 Key Organics / BIONET

1-methyl-4-[5-methyl-4-(2-phenylethyl)-4H-1,2,4-triazol-3-yl]-1H-pyrazol-5-amine

  • MDL number MFCD03305568
  • Cas Number : 329901-23-7
  • Molecular Weight : 282.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69436

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0296_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart