KEY465194488 Key Organics / BIONET

3-(benzylsulfanyl)-1-(4-chlorophenyl)-5,6,7,8-tetrahydroisoquinoline-4-carbonitrile

  • MDL number MFCD03012112
  • Cas Number : 439096-90-9
  • Molecular Weight : 390.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69469

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0353_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart