KEY465194497 Key Organics / BIONET

N-{7-oxo-2H,5H,6H,7H-indeno[5,6-d][1,3]dioxol-5-yl}thiophene-2-carboxamide

  • MDL number MFCD03012252
  • Cas Number : 439097-04-8
  • Molecular Weight : 301.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69495

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-0626_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart