KEY465194500 Key Organics / BIONET

2,2,2-trifluoro-N-(4-methoxy-3-oxo-2,3-dihydro-1H-inden-1-yl)acetamide

  • MDL number MFCD03102804
  • Cas Number : 861209-50-9
  • Molecular Weight : 273.211

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69504

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0642_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart