KEY465194504 Key Organics / BIONET

6-[(5-chloro-1,3-thiazol-2-yl)methyl]-5,7-dimethyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

  • MDL number MFCD03305592
  • Cas Number : 861209-53-2
  • Molecular Weight : 294.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69512

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-0804_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart