KEY465194507 Key Organics / BIONET

N-(7-aminoheptyl)-2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]acetamide

  • MDL number MFCD08281840
  • Cas Number : 882747-18-4
  • Molecular Weight : 351.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69524

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0822_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart