KEY465194509 Key Organics / BIONET

methyl 2-{7-hydroxy-[1,2,4]triazolo[1,5-a]pyrimidin-5-yl}acetate

  • MDL number MFCD03012259
  • Cas Number : 110260-69-0
  • Molecular Weight : 208.177

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69526

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-0824_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart