KEY465194519 Key Organics / BIONET

4-(4-fluorophenyl)-1-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]-1H-pyrazol-5-amine

  • MDL number MFCD03305624
  • Cas Number : 955966-79-7
  • Molecular Weight : 366.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69587

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0922_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart