KEY465194527 Key Organics / BIONET

2,2,2-trichloro-N-[4-(2-phenylethynyl)phenyl]acetamide

  • MDL number MFCD03012882
  • Cas Number : 439097-38-8
  • Molecular Weight : 338.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69600

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-1132_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart