KEY465194529 Key Organics / BIONET

N-[3-(hex-1-yn-1-yl)phenyl]thiophene-2-carboxamide

  • MDL number MFCD04124943
  • Cas Number : 861209-77-0
  • Molecular Weight : 283.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69602

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-1135_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart