KEY465194530 Key Organics / BIONET

3-chloro-N-[3-(hex-1-yn-1-yl)phenyl]benzamide

  • MDL number MFCD04124944
  • Cas Number : 861209-78-1
  • Molecular Weight : 311.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69603

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-1136_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart