KEY465194532 Key Organics / BIONET

N-[4-(hex-1-yn-1-yl)phenyl]-1-phenylmethanesulfonamide

  • MDL number MFCD03012885
  • Cas Number : 439097-41-3
  • Molecular Weight : 327.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69608

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-1141_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart