KEY465194534 Key Organics / BIONET

5-[(4-fluorophenyl)sulfanyl]-1,3-diphenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD03012892
  • Cas Number : 955964-28-0
  • Molecular Weight : 374.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69618

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-1152_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart