KEY465194537 Key Organics / BIONET

1,3-dimethyl 2-[5-(4-tert-butylbenzamido)pyridin-2-yl]propanedioate

  • MDL number MFCD03617950
  • Cas Number : 861209-83-8
  • Molecular Weight : 384.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69629

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart