KEY465194543 Key Organics / BIONET

2-chloro-5-[2-(2-nitrophenyl)ethyl]-1,3-thiazole

  • MDL number MFCD03848405
  • Cas Number : 861209-92-9
  • Molecular Weight : 268.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69644

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2W-0039_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart