KEY465194548 Key Organics / BIONET

5,7,7-trimethyl-6,7-dihydro-1H-1,4-diazepine-2,3-dicarbonitrile

  • MDL number MFCD03618028
  • Cas Number : 51802-58-5
  • Molecular Weight : 188.234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69671

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2W-0256_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart