KEY465194559 Key Organics / BIONET

(2E)-2-[(E)-4-chlorobenzoyl]-3-[3-(pyrimidin-2-yloxy)phenyl]prop-2-enenitrile

  • MDL number MFCD03617967
  • Cas Number : 861210-22-2
  • Molecular Weight : 361.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69703

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2W-0310_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart