KEY465194563 Key Organics / BIONET

2-(3,4-dichlorophenyl)-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one

  • MDL number MFCD03848420
  • Cas Number : 861210-29-9
  • Molecular Weight : 287.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69715

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0340_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart