KEY465194575 Key Organics / BIONET

4,5-dimethyl 1-[2-(methoxycarbonyl)thiophen-3-yl]-1H-1,2,3-triazole-4,5-dicarboxylate

  • MDL number MFCD03787557
  • Cas Number : 861210-49-3
  • Molecular Weight : 325.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69749

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2W-0822_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart