KEY465194578 Key Organics / BIONET

2-nitro-6H,7H,8H,9H,10H,12H-azepino[2,1-b]quinazolin-12-one

  • MDL number MFCD00994584
  • Cas Number : 61938-71-4
  • Molecular Weight : 259.265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69765

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2W-0842_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart