KEY465194580 Key Organics / BIONET

(2E)-3-(dimethylamino)-1-[4-(morpholin-4-yl)phenyl]prop-2-en-1-one

  • MDL number MFCD03617857
  • Cas Number : 1345865-34-0
  • Molecular Weight : 260.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69773

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0854_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart