KEY465194584 Key Organics / BIONET

1-(2-methyl-1,3-benzoxazol-6-yl)-3-(prop-2-en-1-yl)thiourea

  • MDL number MFCD03618038
  • Cas Number : 861210-67-5
  • Molecular Weight : 247.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69780

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2W-0861_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart