KEY465194585 Key Organics / BIONET

4-[(2-chloro-1,3-thiazol-5-yl)methyl]-2-methyl-4,5-dihydro-1,3,4-benzoxadiazepin-5-one

  • MDL number MFCD03848432
  • Cas Number : 861210-68-6
  • Molecular Weight : 307.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69781

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0864_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart