KEY465194586 Key Organics / BIONET

ethyl 2-(2-methyl-5-oxo-4,5-dihydro-1,3,4-benzoxadiazepin-4-yl)acetate

  • MDL number MFCD03848433
  • Cas Number : 861210-69-7
  • Molecular Weight : 262.265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69782

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0866_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart