KEY465194590 Key Organics / BIONET

2-({1-[4-(thiophen-2-yl)pyrimidin-2-yl]-1H-pyrrol-2-yl}methylidene)propanedinitrile

  • MDL number MFCD03848436
  • Cas Number : 861210-74-4
  • Molecular Weight : 303.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69788

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0872_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart